Learn More
Electron Microscopy Sciences Branson Ultra Sonic Bath Model 8800-DTH, 115 V
SDP
Catalog No. 501899438
Encompass_Preferred
With a unique sweep frequency tech to eliminate standing waves, & a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry std for quality, reliability, & precision cleaning.Constant activity/power tracking auto maintains the ultrasonic power by adjusting for changes in liquid level & temp caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.Temperature settings up to 69°C/156.2°F can be set in Celsius or Fahrenheit & programmed through the front panel for easy access & monitoring. High-temperature visual alarm/auto shut-off provides added safety. Ultrasonic Cleaner w/ digital timer, heater degas, & temp monitor, tank size: 19.5inx11inx6in, tank capacity: 5.5 gal, drain: yes, overall size: 24inx18inx14.5in.
Catalog No. 50-189-9438 Supplier Electron Microscopy Sciences Supplier No. 71040DTH
With a unique sweep frequency tech to eliminate standing waves, & a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry std for quality, reliability, & precision cleaning.Constant activity/power tracking auto maintains the ultrasonic power by adjusting for changes in liquid level & temp caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.Temperature settings up to 69°C/156.2°F can be set in Celsius or Fahrenheit & programmed through the front panel for easy access & monitoring. High-temperature visual alarm/auto shut-off provides added safety. Ultrasonic Cleaner w/ digital timer, heater degas, & temp monitor, tank size: 19.5inx11inx6in, tank capacity: 5.5 gal, drain: yes, overall size: 24inx18inx14.5in.