Learn More
STONYLAB Ultrasonic Cleaner, 2 Liter, Digital Heating & Timer, 40 kHz Lab Glassware Cleaner, 304 Stainless Steel, 110volt
Catalog No. NC4035846
Encompass
Product Overview: stonyLab Ultrasonic Cleaner is professional designed to provide effective and efficient cleaning of laboratory glassware and instruments with simply, user-friendly operationDisplay: clearly, intuitively display temperature and time with two digital displays, ensuring precise cleaning temperature and time control; heating could reduce the adhesion of stains and boost the cleaning powerSpecifications: operation frequency: 40 kHz; temperature is adjustable up to 80°C (176℉); the timer can be set from 1-99 minutesMaterial: made of 304 stainless steel, our safe, reliable ultrasonic cleaner is corrosion and high temperature resistant; basket helps reduce friction for small parts that need cleaning
Catalog No. NC4035846 Supplier STONYLAB Supplier No. SL000091
Product Overview: stonyLab Ultrasonic Cleaner is professional designed to provide effective and efficient cleaning of laboratory glassware and instruments with simply, user-friendly operationDisplay: clearly, intuitively display temperature and time with two digital displays, ensuring precise cleaning temperature and time control; heating could reduce the adhesion of stains and boost the cleaning powerSpecifications: operation frequency: 40 kHz; temperature is adjustable up to 80°C (176℉); the timer can be set from 1-99 minutesMaterial: made of 304 stainless steel, our safe, reliable ultrasonic cleaner is corrosion and high temperature resistant; basket helps reduce friction for small parts that need cleaning