Learn More
Roboz Surgical Roboz Sentry Ultrasonic Cleaner, 2500 mL, Stainless Steel, 1 ea
SDP
Catalog No. NC9081455
Encompass
This high-power ultrasonic cleaner utilizes advanced cavitation technology to effectively clean surgical instruments and laboratory tools. By generating micron-sized bubbles that implode with high velocity, the unit reaches into tight spaces and crevices inaccessible by manual cleaning methods. Designed for efficiency, it features a digital control panel with pre-optimized cleaning cycles to ensure thorough results.
  • Features a high-capacity elongated stainless steel tank
  • Offers five preset digital timer cycles
  • Operates at an ultrasonic frequency of 42000 Hz
  • Provides heating capability up to 65 °C
  • Includes a protective plastic tray and power cord
  • Supports AC100-120V power input
  • Designed for quiet operation with a compact footprint
Catalog No. NC9081455 Supplier Roboz Surgical Supplier No. RS9911
This high-power ultrasonic cleaner utilizes advanced cavitation technology to effectively clean surgical instruments and laboratory tools. By generating micron-sized bubbles that implode with high velocity, the unit reaches into tight spaces and crevices inaccessible by manual cleaning methods. Designed for efficiency, it features a digital control panel with pre-optimized cleaning cycles to ensure thorough results.
  • Features a high-capacity elongated stainless steel tank
  • Offers five preset digital timer cycles
  • Operates at an ultrasonic frequency of 42000 Hz
  • Provides heating capability up to 65 °C
  • Includes a protective plastic tray and power cord
  • Supports AC100-120V power input
  • Designed for quiet operation with a compact footprint