Learn More
MSC REED Instruments Ultrasonic Couplant Gel, Water Soluble, Slow Drying, 250 mL
Catalog No. 5000513167
Encompass
This ultrasonic couplant gel is designed to provide effective acoustic transmission for ultrasonic thickness measurement applications. The water-soluble formulation offers a consistent, slow-drying medium that ensures optimal contact between the transducer and the testing surface.
  • Water soluble for easy cleanup
  • Slow-drying formula for extended testing time
  • Provides good transducer lubrication
  • Includes an 8.45 fl oz (250 mL) applicator bottle
Catalog No. 50-005-13167 Supplier MSC Supplier No. 10165710
This ultrasonic couplant gel is designed to provide effective acoustic transmission for ultrasonic thickness measurement applications. The water-soluble formulation offers a consistent, slow-drying medium that ensures optimal contact between the transducer and the testing surface.
  • Water soluble for easy cleanup
  • Slow-drying formula for extended testing time
  • Provides good transducer lubrication
  • Includes an 8.45 fl oz (250 mL) applicator bottle