Learn More
Darwin Chambers Company ULTRASONIC HUMIDIFICATION SYS
SDP
Catalog No. NC1531286
Encompass_Preferred
Option to add Microprocessor-control of ultrasonic humidification system, ambient to 90%, not to exceed a 40°C dew-point. Provides control of ±0.2% at sensor and display resolution of ±0.1%. Our exclusive solid state controlled ultrasonic humidification method generates a cool mist that once introduced into the chamber, eliminates hot spots normally associated with steam-based humidification systems. It greatly reduces required operational and maintenance costs as compared to traditional humidification systems.
Catalog No. NC1531286 Supplier Darwin Chambers Company Supplier No. HUM2
Option to add Microprocessor-control of ultrasonic humidification system, ambient to 90%, not to exceed a 40°C dew-point. Provides control of ±0.2% at sensor and display resolution of ±0.1%. Our exclusive solid state controlled ultrasonic humidification method generates a cool mist that once introduced into the chamber, eliminates hot spots normally associated with steam-based humidification systems. It greatly reduces required operational and maintenance costs as compared to traditional humidification systems.