Learn More
Electron Microscopy Sciences Branson Ultrasonic Model B200, 110 V
SDP
Catalog No. 50930731
Encompass_Preferred
A small economic unit for precise cleaning in a wide variety of applications in the lab. Comes with a 15 ounce stainless steel tank, basket and cover. It features a 5 minute timer which shuts off automatically. Comes complete with parts holder. 115 volt; 50/60V
Catalog No. 50-930-731 Supplier Electron Microscopy Sciences Supplier No. 71048
A small economic unit for precise cleaning in a wide variety of applications in the lab. Comes with a 15 ounce stainless steel tank, basket and cover. It features a 5 minute timer which shuts off automatically. Comes complete with parts holder. 115 volt; 50/60V