Learn More
Electron Microscopy Sciences Branson Ultrasonic Model B200, 220 V
SDP
Catalog No. 50930732
Encompass_Preferred
A small economic unit for precise cleaning in a wide variety of applications in the lab. Comes with a 15 ounce stainless steel tank, basket and cover. It features a 5 minute timer which shuts off automatically. Comes complete with parts holder. 115 volt; 50/60V
Catalog No. 50-930-732 Supplier Electron Microscopy Sciences Supplier No. 7104822
A small economic unit for precise cleaning in a wide variety of applications in the lab. Comes with a 15 ounce stainless steel tank, basket and cover. It features a 5 minute timer which shuts off automatically. Comes complete with parts holder. 115 volt; 50/60V