Learn More
Sigma Aldrich Fine Chemicals Biosciences Ultrasonic processor 130 W1EA
Catalog No. 502909054
Encompass
.nbsp20 kHz.nbspDigital wattmeter.nbspFull function pulser off cycle 1s on cycle variable from 1 to 10s.nbspElectronic timer variable from 1 to 10 hour..nbspVariable output control.nbspAutomatic tuningFor processing medium volume samples from 150 muL to 150 mL. Unit has a piezoelectric converter and a footswitch connector. Shipped ready for operation with titanium probe 138 mm L x 6 mm tip diam. tool kit and instruction manual.
Catalog No. 50-290-9054 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4126001EA
.nbsp20 kHz.nbspDigital wattmeter.nbspFull function pulser off cycle 1s on cycle variable from 1 to 10s.nbspElectronic timer variable from 1 to 10 hour..nbspVariable output control.nbspAutomatic tuningFor processing medium volume samples from 150 muL to 150 mL. Unit has a piezoelectric converter and a footswitch connector. Shipped ready for operation with titanium probe 138 mm L x 6 mm tip diam. tool kit and instruction manual.