Learn More
Sigma Aldrich Fine Chemicals Biosciences Ultrasonic processors 500W1EA
Catalog No. 502908645
Encompass
20 kHz units are excellent for processing samples from 250 muL to several liters on a flow-through basis. Order calibrated SS temperature probe separately..nbspAutomatic tuning and frequency control.nbspIntegrated temperature controller from 1C to 100C.nbspStores up to 10 procedures.nbspReal time display.nbspControl process time from 1 second to 10 hours.nbspShipped complete with a 1/2 in. (13 mm) probe (630-0220) with replaceable tip tool kit and instruction manualApplications Cell disruption in bacteria spores and tissues accelerating catalytic reactions extractions of serums toxins enzymes and viruses from organic sources solubilizing difficult compounds emulsification to 0.01 mm homogenizing liquids sample preparation from microscopy and particle size analysis.
Catalog No. 50-290-8645 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5114711EA
Add to Cart
Add to Cart
Request a Quote
20 kHz units are excellent for processing samples from 250 muL to several liters on a flow-through basis. Order calibrated SS temperature probe separately..nbspAutomatic tuning and frequency control.nbspIntegrated temperature controller from 1C to 100C.nbspStores up to 10 procedures.nbspReal time display.nbspControl process time from 1 second to 10 hours.nbspShipped complete with a 1/2 in. (13 mm) probe (630-0220) with replaceable tip tool kit and instruction manualApplications Cell disruption in bacteria spores and tissues accelerating catalytic reactions extractions of serums toxins enzymes and viruses from organic sources solubilizing difficult compounds emulsification to 0.01 mm homogenizing liquids sample preparation from microscopy and particle size analysis.