Learn More
Valutek™ Ultrasonic seal Standard Weight Polyester Wiper
Catalog No. 191502002
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
Catalog No. Dimensions (L x W)
19-150-2002 9 x 9 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-2002 Supplier Valutek™ Supplier No. VTPNWUS99
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed from 100% continuous filament polyester in a double-knit, no-run, interlock pattern with an ultrasonic sealed edge. This is an ultraclean and highly absorbent wiper with extremely low levels of particulate and extractable counts.
  • 100% continuous-filament double-knit polyester fiber
  • Ultrasonic sealed edge for reduced fiber contamination
  • Moderate absorbency and abrasion resistance
  • Excellent chemical compatibility with IPA and other common solvents
  • Extremely low levels of particulate and extractable counts
  • Laundered and packaged in a cleanroom
  • All Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 1 -10 (ISO 3-4) critical environment
  • It is designed for critical processing applications where the highest level of contamination control is required
  • It is also commonly used for cleaning medical device products; applying, removing, cleaning and disinfecting solutions; is steam autoclavable for aseptic environments; and used for flat panel, optics cleaning, and chips multi-processing (CMP)
Specifications
Material Polyester
Dimensions (L x W) 9 x 9 in.
Sterility Non-sterile