Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Ultrasonic Sensor 1 In NPT 24

Supplier:  Grainger 1AAA7

Encompass_Preferred

Ultrasonic Sensor 1 In NPT 24 VDC

Catalog No. 50-279-4375


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.