Learn More
World Precision Instrument Alconox Concentrated Cleaning Powder for Surgical Instruments
SDP
Catalog No. 50822190
Encompass
A safe, concetrated cleaner perfect for surgical instruments and labware
Alconox is a concentrated, anionic detergent for manual and ultrasonic cleaning. This detergent gives you reliable results without interfering residues, because it rinses away easily. Alconox cleaner is Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. It is an excellent replacement for corrosive acids and hazardous solvents.
Catalog No. 50-822-190 Supplier World Precision Instrument Supplier No. 13740
A safe, concetrated cleaner perfect for surgical instruments and labware
Alconox is a concentrated, anionic detergent for manual and ultrasonic cleaning. This detergent gives you reliable results without interfering residues, because it rinses away easily. Alconox cleaner is Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. It is an excellent replacement for corrosive acids and hazardous solvents.