Learn More
ORS Nasco ULTRASPEC 2000 ADJ CL XTR 1PR
Catalog No. 502551590
Encompass_Preferred
Honeywell part# S0250X. Honeywell Uvex Ultraspec 2000 Safety Glasses Lens Type: Anti-fog Lens Tint: Clear Lens Material: Polycarbonate Recommended Applications: Safety Temple Style: Adjustable-length; spatula
Catalog No. 50-255-1590 Supplier ORS Nasco Supplier No. 763S0250X
Honeywell part# S0250X. Honeywell Uvex Ultraspec 2000 Safety Glasses Lens Type: Anti-fog Lens Tint: Clear Lens Material: Polycarbonate Recommended Applications: Safety Temple Style: Adjustable-length; spatula