Learn More
Waters Corp Ultrastyragel GPC Column, 103 Å, 19 mm x 300 mm, THF
Catalog No. 50785232 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Ultrastyragel GPC Columns are designed for gel permeation chromatography, offering precise separation based on molecular size.
  • Column with 103 Å pore size.
  • Dimensions of 19 mm I.D. x 300 mm.
  • Recommended for use with tetrahydrofuran (THF) as a mobile phase.
  • Effective molecular weight range of 200 to 30,000.
Catalog No. 50-785-232 Supplier Waters Corp Supplier No. WAT025861
Ultrastyragel GPC Columns are designed for gel permeation chromatography, offering precise separation based on molecular size.
  • Column with 103 Å pore size.
  • Dimensions of 19 mm I.D. x 300 mm.
  • Recommended for use with tetrahydrofuran (THF) as a mobile phase.
  • Effective molecular weight range of 200 to 30,000.