Learn More
Waters Corp Waters Ultrastyragel GPC Column, 10000 Å, 5 μm, 19 × 300 mm
Supplier: Waters Corp WAT025862
This Ultrastyragel GPC column is designed for high-resolution preparative Gel Permeation Chromatography (GPC) applications. It features a 10000 Å pore size and 5 μm spherical particles, suitable for separating molecules from 5,000 to 600,000 molecular weight in Tetrahydrofuran (THF) solvent. Column dimensions are 19 × 300 mm.
- High-resolution preparative GPC applications
- 10000 Å pore size
- 5 μm particle size
- Spherical particle shape
- 19 mm inner diameter
- 300 mm length
- 5,000 to 600,000 molecular weight range
- Tetrahydrofuran (THF) packing solvent
- pH range: 2 to 12
- Size Exclusion Chromatography (SEC) technique
- HPLC system compatible
- 1 unit per package
- Contains hazardous material requiring special freight handling and additional charges
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.