Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Ultrastyragel GPC Column, 100000Å, 5 μm, 19 mm x 300 mm

Catalog No. 50785241 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Ultrastyragel GPC Column is designed for high-resolution preparative Gel Permeation Chromatography (GPC) applications.

  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range of 2 to 12
  • Molecular weight range of 50,000 to 4,000,000
  • Spherical particle shape
  • Particle size of 5 μm
  • Pore size of 100,000 Å
  • Column format
  • Toluene packing solvent
  • HPLC system compatibility
  • SEC technique
  • Inner diameter of 19 mm
  • Length of 300 mm
  • 1 unit per package
  • Contains hazardous material requiring special freight handling

Catalog No. 50-785-241 Supplier Waters Corp Supplier No. WAT025870
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Ultrastyragel GPC Column is designed for high-resolution preparative Gel Permeation Chromatography (GPC) applications.

  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range of 2 to 12
  • Molecular weight range of 50,000 to 4,000,000
  • Spherical particle shape
  • Particle size of 5 μm
  • Pore size of 100,000 Å
  • Column format
  • Toluene packing solvent
  • HPLC system compatibility
  • SEC technique
  • Inner diameter of 19 mm
  • Length of 300 mm
  • 1 unit per package
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.