Learn More
Waters Corp Waters HPLC Column, 10^6 Angstrom Pore, 5 um Particle, 19 mm x 300 mm
Catalog No. 50785235 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC column is designed for high resolution preparative GPC applications. It provides high-efficiency GPC separations for isolation and sample cleanup, enabling complex separations to be performed on a single, more efficient preparative column.
  • Provides high-efficiency GPC separations for isolation and sample cleanup
  • Enables performing separations that previously required multiple smaller columns on a single, more efficient preparative column
  • Features a 106Å pore size
  • Equipped with 5.0 μm particles
  • Measures 19 mm inner diameter and 300 mm length
  • Offers an effective molecular weight range of 200,000 to 10,000,000
  • Supports a pH range of 2 to 12
Catalog No. 50-785-235 Supplier Waters Corp Supplier No. WAT025864
This HPLC column is designed for high resolution preparative GPC applications. It provides high-efficiency GPC separations for isolation and sample cleanup, enabling complex separations to be performed on a single, more efficient preparative column.
  • Provides high-efficiency GPC separations for isolation and sample cleanup
  • Enables performing separations that previously required multiple smaller columns on a single, more efficient preparative column
  • Features a 106Å pore size
  • Equipped with 5.0 μm particles
  • Measures 19 mm inner diameter and 300 mm length
  • Offers an effective molecular weight range of 200,000 to 10,000,000
  • Supports a pH range of 2 to 12