Learn More
Waters Corp Waters GPC Column, 100 Å, 5 μm, 7.8 × 300 mm, THF
Catalog No. 50784975 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These GPC columns are designed for high-resolution, low molecular weight applications. They provide high-efficiency GPC separations for compound isolation and sample clean-up of water-soluble polymers. The column is commonly used for characterizing oligomers and biological molecules such as polysaccharides, nucleic acids, proteins, and peptides, improving separation speed and reducing solvent consumption.
  • High resolution
  • Low adsorption
  • Wide molecular weight range: 50 to 1500
  • Operates within a pH range of 2 to 12
  • Polymer particle substrate
  • Spherical particle shape
  • Packing solvent: THF
  • Suitable for HPLC systems
  • Manufactured with stringent quality control
Catalog No. 50-784-975 Supplier Waters Corp Supplier No. WAT010570
These GPC columns are designed for high-resolution, low molecular weight applications. They provide high-efficiency GPC separations for compound isolation and sample clean-up of water-soluble polymers. The column is commonly used for characterizing oligomers and biological molecules such as polysaccharides, nucleic acids, proteins, and peptides, improving separation speed and reducing solvent consumption.
  • High resolution
  • Low adsorption
  • Wide molecular weight range: 50 to 1500
  • Operates within a pH range of 2 to 12
  • Polymer particle substrate
  • Spherical particle shape
  • Packing solvent: THF
  • Suitable for HPLC systems
  • Manufactured with stringent quality control