Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Ultrastyragel Column, 5 μm, 100 Å, 19 mm x 300 mm

Catalog No. 50785230 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column is designed for high resolution preparative GPC applications.

  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range: 2 to 12
  • Molecular weight range: 50 to 1500
  • Spherical particle shape
  • 5 μm particle size
  • 100 Å pore size
  • Column format
  • THF packing solvent
  • HPLC system
  • SEC technique
  • 19 mm inner diameter
  • 300 mm length
  • 1 unit per package

Catalog No. 50-785-230 Supplier Waters Corp Supplier No. WAT025859
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This column is designed for high resolution preparative GPC applications.

  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range: 2 to 12
  • Molecular weight range: 50 to 1500
  • Spherical particle shape
  • 5 μm particle size
  • 100 Å pore size
  • Column format
  • THF packing solvent
  • HPLC system
  • SEC technique
  • 19 mm inner diameter
  • 300 mm length
  • 1 unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.