Learn More
Waters Corp Ultrastyragel Column, 100 Å, 5 μm, 19 mm × 300 mm, Toluene
Catalog No. 50785237 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Ultrastyragel Preparative Column provides high-efficiency GPC separations for isolation and sample cleanup. It enables separations that typically require multiple smaller columns to be performed on a single, more efficient preparative column.
  • Designed for high-efficiency GPC separations
  • Ideal for isolation and sample cleanup
  • Features a 100 Å pore size
  • Suitable for molecular weight range of 50 to 1500
  • Optimized for use with Toluene as a solvent
  • Dimensions are 19 mm I.D. × 300 mm
Catalog No. 50-785-237 Supplier Waters Corp Supplier No. WAT025866
This Ultrastyragel Preparative Column provides high-efficiency GPC separations for isolation and sample cleanup. It enables separations that typically require multiple smaller columns to be performed on a single, more efficient preparative column.
  • Designed for high-efficiency GPC separations
  • Ideal for isolation and sample cleanup
  • Features a 100 Å pore size
  • Suitable for molecular weight range of 50 to 1500
  • Optimized for use with Toluene as a solvent
  • Dimensions are 19 mm I.D. × 300 mm