Learn More
Waters Corp Waters Corporation Column, 500 Å, 5 μm, 7.8 X 300 mm
Supplier:  Waters Corp WAT010571
Encompass_Preferred
These high resolution GPC columns are designed for low molecular weight applications. They offer key analytical capabilities including a wide range of molecular weight separations, low adsorption, and excellent resolution. These columns are built to increase efficiency in plates and/or meters by two to three times, accelerating separation while lowering solvent consumption for preparative isolation.
  • High resolution for low molecular weight applications
  • Wide range of molecular weight separations
  • Low adsorption
  • Excellent resolution
  • Increased efficiency in plates and/or meters, leading to faster separation
  • Lower solvent consumption for preparative isolation
  • Can perform separations that previously required multiple smaller columns
  • Available in a variety of chemistries, lengths, pore designs, and particle sizes
Catalog No. 50-784-976