Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, 500 Å, 5 μm, 19 mm x 300 mm, Toluene

Catalog No. 50785238 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column is designed for high resolution preparative GPC applications. It features a spherical polymer particle substrate and is packed in Toluene, supporting a molecular weight range from 100 to 10000.

  • Spherical polymer particle substrate
  • Particle size of 5 μm
  • Pore size of 500 Å
  • Inner diameter of 19 mm
  • Length of 300 mm
  • Packed in Toluene
  • Molecular weight range from 100 to 10000
  • Suitable for SEC/GPC separation mode
  • Compatible with HPLC systems
  • pH range from 2 to 12

Catalog No. 50-785-238 Supplier Waters Corp Supplier No. WAT025867
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This column is designed for high resolution preparative GPC applications. It features a spherical polymer particle substrate and is packed in Toluene, supporting a molecular weight range from 100 to 10000.

  • Spherical polymer particle substrate
  • Particle size of 5 μm
  • Pore size of 500 Å
  • Inner diameter of 19 mm
  • Length of 300 mm
  • Packed in Toluene
  • Molecular weight range from 100 to 10000
  • Suitable for SEC/GPC separation mode
  • Compatible with HPLC systems
  • pH range from 2 to 12

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.