Learn More
Waters Corp Waters Ultrastyragel Column, 5 μm, 19 mm X 300 mm, GPC
Catalog No. 50785236 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Ultrastyragel Linear Column is designed for high-resolution preparative Gel Permeation Chromatography (GPC) applications. This column features 5 μm spherical polymer particles and dimensions of 19 mm inner diameter by 300 mm length. It is compatible with THF as a mobile phase and is suitable for separating molecules with a molecular weight range of 2,000 to 4,000,000. It operates within a pH range of 2 to 12.
  • High-resolution preparative GPC applications.
  • Spherical polymer particles, 5 μm.
  • Dimensions of 19 mm inner diameter by 300 mm length.
  • Compatible with THF mobile phase.
  • Suitable for molecular weights from 2,000 to 4,000,000.
  • Operates within a pH range of 2 to 12.
Catalog No. 50-785-236 Supplier Waters Corp Supplier No. WAT025865
The Ultrastyragel Linear Column is designed for high-resolution preparative Gel Permeation Chromatography (GPC) applications. This column features 5 μm spherical polymer particles and dimensions of 19 mm inner diameter by 300 mm length. It is compatible with THF as a mobile phase and is suitable for separating molecules with a molecular weight range of 2,000 to 4,000,000. It operates within a pH range of 2 to 12.
  • High-resolution preparative GPC applications.
  • Spherical polymer particles, 5 μm.
  • Dimensions of 19 mm inner diameter by 300 mm length.
  • Compatible with THF mobile phase.
  • Suitable for molecular weights from 2,000 to 4,000,000.
  • Operates within a pH range of 2 to 12.