Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UltraTape™ 1110 Polyethylene Medium-High Adhesion
SDP

Catalog No. 191202447 Shop All Ultra Tape Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-120-2447 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-120-2447 Supplier UltraTape™ Supplier No. 1110WH100P3D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Adhesive Type Acrylic
Color White
For Use With (Application) Semiconductor
Tensile Strength 12 lb./in.
Thickness (English) 5.5 mil ±.7 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.