Learn More
UltraTape™ 1154 Polyethylene Medium Adhesion Tape
SDP
Catalog No. 19168625 Shop All Ultra Tape Products
Change view
Click to view available options
Thickness (Metric):
1.5 mil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (Metric)
19-168-625 1.5 mil
19-168-626 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-168-625 Supplier UltraTape™ Supplier No. 1154WH150P3D
Specifications
Color White
Tensile Strength 18 lb./in.
Thickness (English) 1.5 mil
Thickness (Metric) 1.5 mil
Length (English) 36 yd.