Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UltraTape™ UltraClean Polyethylene Medium Adhesion Tape
SDP

Catalog No. 19168972 Shop All Ultra Tape Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-168-972 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-168-972 Supplier UltraTape™ Supplier No. 1112CL100P3D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Adhesive Type Acrylic
Color Clear
Material Polyethylene
Product Type Cleanroom Packing Tape
For Use With (Application) For cleanroom packing, wafer shipping boxes, bags
Dimensions (L x W) 1 in. x 36 yd.
Length (English) 36 yd.
Width (English) 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.