Learn More
MSC UltraTape Vinyl Medium Adhesion, 1.5x36 yd, White, 1 ea
Catalog No. 5000795764
Encompass
UltraTape Vinyl Medium Adhesion tape is designed for use in controlled environments. This tape provides reliable adhesion while being easy to remove, making it suitable for temporary securing and marking tasks in cleanrooms.
  • Provides medium adhesion for secure but temporary holding
  • Leaves no residue upon removal
  • Durable vinyl construction resists tears and punctures
  • Suitable for various cleanroom applications
  • White color for clear visibility
Catalog No. 50-007-95764 Supplier MSC Supplier No. 20446902
UltraTape Vinyl Medium Adhesion tape is designed for use in controlled environments. This tape provides reliable adhesion while being easy to remove, making it suitable for temporary securing and marking tasks in cleanrooms.
  • Provides medium adhesion for secure but temporary holding
  • Leaves no residue upon removal
  • Durable vinyl construction resists tears and punctures
  • Suitable for various cleanroom applications
  • White color for clear visibility