Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety UltraTech™ CutPro™ Nitrile Foam Coated Mechanics Gloves with TPR Padding

Catalog No. 19812723 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-723 2X-Large
19-812-721 Small
19-812-722 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-812-723 Supplier MCR Safety Supplier No. UT2952XXL
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves feature a 13 gauge HPPE/synthetic knit shell with a foam nitrile coating and TPR padding protection against cuts and abrasions.

  • 13-gauge HPPE/synthetic knit shell is comfortable and flexible
  • Foam nitrile coated palm and fingers enhances grip capabilities
  • TPR padding provides impact protection for the back of the hand
  • ID panel on the wrist for user personalization
  • Touch screen friendly

Specifications

ANSI Abrasion Rating ANSI 5
Back Material TPR Padding
Coating Material Nitrile Foam
Color Gray, Orange
Cut Resistance Rating ANSI A4
Disposable Reusable
Grade Industrial
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanic's Gloves
Size 2X-Large
Finger Style Full
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 4
Series UT2952
Yarn Gauge 13
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.