Learn More
Youngstown Barrel & Drum Ultratech Ultra-Drain Guard
Description
- Oil and sediment model removes up to 0.87 gal. of hydrocarbons, dirt, sand and other contaminants
- For use in industrial facilities, construction sites, and parking lots
- Removes contaminants like litter, sand, and oil from stormwater flow
- Made from geotextile fabric
- Easy to install
Specifications
Specifications
| Material | Geotextile |
| Type | Oil and Sediment Drain Guard |
| For Use With (Application) | Removes up to 0.87 gal. of hydrocarbons, dirt, sand and other contaminants |
| For Use With (Equipment) | For use in industrial facilities, construction sites, and parking lots |