Learn More
Youngstown Barrel & Drum Ultratech Ultra-Hard Top P2 Spill Pallet™

Description
- Provides storage for up to two, 55-gallon drums
- Made from polyethylene with heavy-duty steel cover lift assembly
- Removable deck grates and rugged steel brackets
- Drums positioned on deck above sump area, elevated from any spilled or leaked materials
- UV inhibitor in PE resin resists degrading effects of sunlight
- 66-gallon containment capacity meets SPCC and EPA Spill Containment Regulations
Specifications
Specifications
| Capacity (Metric) | 250 L |
| Material | Polyethylene |
| Type | Ultra-Hard Top™ P2 Spill Pallet |
| For Use With (Application) | Storage of virgin chemicals, drum dispensing, hazardous and satellite waste collection, storage of spill response materials |
| Includes | Heavy-duty steel cover lift. assembly |
| Model | Two-Drum Hard Top Spill Pallet without Drain |
| Resistant | UV, degrading effects of sunlight |
| Dimensions (L x W x H) | 1448 x 775 x 1613 mm |
| Weight (English) | 173 lb. |
| Weight (Metric) | 78.5 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.