Learn More
Scytek Laboratories ULTRATEK ANTI-POLYVALENT,500ML
SDP
Catalog No. NC1835348
Encompass_Preferred
The UltraTek line a leading edge system, designed to provide optimal staining with incubation times of 10 minutes each for the link antibody and enzyme label. For most procedures, commercially available primary antibodies can be diluted up to 50% further than with other systems.
Catalog No. NC1835348 Supplier Scytek Laboratories Supplier No. ABN500
The UltraTek line a leading edge system, designed to provide optimal staining with incubation times of 10 minutes each for the link antibody and enzyme label. For most procedures, commercially available primary antibodies can be diluted up to 50% further than with other systems.