Learn More
Scytek Laboratories ULTRATEK HORSERADISH PEROXIDAS
SDP
Catalog No. NC1835350
Encompass_Preferred
UltraTek Horseradish Peroxidase,500MLThe UltraTek line is a leading edge system, designed to provide optimal staining with incubation times of 10 minutes each for the link antibody and enzyme label. For most procedures, commercially available primary antibodies can be diluted up to 50% further than with other systems.
Catalog No. NC1835350 Supplier Scytek Laboratories Supplier No. ABL500
UltraTek Horseradish Peroxidase,500MLThe UltraTek line is a leading edge system, designed to provide optimal staining with incubation times of 10 minutes each for the link antibody and enzyme label. For most procedures, commercially available primary antibodies can be diluted up to 50% further than with other systems.