Learn More
Ted Pella Inc ULTRATHIN QFOIL 1.2UM 100GRIDS
SDP
Catalog No. NC1811330
Encompass
Ultrathin Carbon on Quantifoil, Orthogonal Array of 1.2µm Diameter Holes - 1.3µm Separation, mounted on a 300M Cu grid, package of 100 grids
Catalog No. NC1811330 Supplier Ted Pella Inc Supplier No. 668300CU100
Add to Cart
Add to Cart
Ultrathin Carbon on Quantifoil, Orthogonal Array of 1.2µm Diameter Holes - 1.3µm Separation, mounted on a 300M Cu grid, package of 100 grids