Learn More
Electron Microscopy Sciences DiATOME Resharpen UltraTrim Wet Room Temp, 3.0 mm
SDP
Catalog No. 501900200
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 3.0 mm
Catalog No. 50-190-0200 Supplier Electron Microscopy Sciences Supplier No. UWTTR
Add to Cart
Add to Cart
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 3.0 mm