Learn More
Electron Microscopy Sciences DiATOME Resharpen UltraTrim Dry Room Temp, 4.0 mm
SDP
Catalog No. 501900197
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 4.0 mm
Catalog No. 50-190-0197 Supplier Electron Microscopy Sciences Supplier No. UTT40R
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 4.0 mm