Learn More
Hach Company Ultraviolet Lamp
Supplier:  Hach Company 2184311
Encompass
Lamp-Ultraviolet-Long-Wave Replacement BulbMid-size (6 Watt) high intensity hand-held and line-operated Intensity at 15 2 cm (6in ) 750 W/cm2 Filters Two 6 x 7 6 cm (2 4 x 3in ) in snap on/off frame Overall dimensions 37 5 x 8 9 x 6 7 cm (14 8 x 3 5 x 2 6in ) Weight 0 91 kg (2 lbs) UL listed Meets existing OSHA electrical standards 365 nm UV Products
Catalog No. 50-002-07074