Learn More
Med Vet International Ultraviolet Wood's Light, 4-watt, 110V AC, J0312Q, Each
Catalog No. 5000585501
Encompass_Preferred
A hand-held lamp with 110V plug-in. High impact plastic housing.Ultraviolet light at wavelength of 365nm with intensity of 180uW/cm2 at 6"75/8" x 27/8" x 2". 1 lb.4-watts
Catalog No. 50-005-85501 Supplier Med Vet International Supplier No. JOR312Q
A hand-held lamp with 110V plug-in. High impact plastic housing.Ultraviolet light at wavelength of 365nm with intensity of 180uW/cm2 at 6"75/8" x 27/8" x 2". 1 lb.4-watts