Learn More
UltraVision™ LifeLocator™ II Life Detection System
Catalog No. 18680201
Change view
Click to view available options
Product Type:
PDA Control Unit
Catalog No. Product Type
18-680-201 PDA Control Unit
1 options
Catalog No. 18-680-201 Supplier UltraVision™ Supplier No. FGPDACULL
Add to Cart
Add to Cart
Includes
Additional batteries, chargers, and transit case. Replacement PDA available.
Surpasses traditional video, audio, and canine rescue methods to improve odds of rescue over recovery
Rugged, weather- and water-resistant PDA control unit is lightweight, portable and completely wireless. Deploys in seconds, allowing large disaster areas to be covered quickly.
  • Detects and locates motion as slight as shallow breathing within 30 seconds
  • Large field of detection—up to 4500 cu. ft. (127m3)
  • Effective through reinforced concrete and other building materials without the need for drilling
  • Deployment takes seconds, not minutes or hours, so large disaster areas can be covered in much less time
  • Handheld PDA control unit displays a black square when motion is detected; displays a red circle when only breathing is detected
  • Indicates approximate distance to the victim from the sensor
  • Unaffected by odors, wind shifts, or other bio-sensitivities that affect canine detection
  • Operates in loud environments; not affected by rescue-area-generated sounds or other noise
  • Firmware can be easily updated over hard-wired or wireless networks
  • Low power requirements; no required maintenance
Certifications
FCC certified on GSA Schedule and Department of Homeland Security's Approved Equipment List
Specifications
Product Type PDA Control Unit
For Use With (Application) LifeLocator II motion detection system
Weight (English) 1.06 lb.
Weight (Metric) 0.48 kg