Learn More
Description
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: None provided
Includes:
TL-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH)
TL-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH)
Note:
UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.
Specifications
Specifications
| Count | 625 to 1250 slides |
| Includes | 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH) |
| Chromagen | None provided |
| Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
| Storage Requirements | Store at 2° to 8°C. Each component is stable for 18 months. |
| Enzyme | Peroxidase |
| Product Line | Lab Vision, UltraVision |
| Substrate | None provided |
| Quantity | 125 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.