Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ UltraVision LP HRP Polymer & DAB Detection System

Catalog No. TL125HD Shop All Epredia Products
Change view
Click to view available options
Quantity:
15 mL
60 mL
125 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL125HD 125 mL
TL015HD 15 mL
TL060HD 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. TL125HD Supplier Epredia™ Supplier No. TL125HD

missing translation for 'validMainframeMsg'

Epredia™ Lab Vision UltraVision LP Detection System: HRP Polymer/DAB Plus Chromogen is a sensitive detection system based on biotin-free, polymer technology.

UltraVision LP is the latest technology in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This second-generation polymer system is composed of smaller polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification.1 Ultimately, this gives the user higher sensitivity and antibody efficiency.2 With UltraVision LP, you use less antibody and obtain better signal-to-noise ratios. UltraVision LP is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: Diaminobenzidine (DAB)

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: Diaminobenzidine (DAB)

Includes:

TL-015-HD: 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL Ultra V Block (TA-015-UB), 15mL Primary Antibody Enhancer (TL-015-PB), 15mL HRP Polymer (TL-015-PH), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX)

TL-060-HD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH), 60mL DAB Plus Substrate (TA-060-HSX), 2mL DAB Plus Chromogen (TA-002-HCX)

TL-125-HD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH), 125mL DAB Plus Substrate (TA-125-HSX), 4mL DAB Plus Chromogen (TA-004-HCX)

Note:

UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.

Specifications

Quantity 125 mL
Count 625 to 1250 slides
Includes 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH), 125mL DAB Plus Substrate (TA-125-HSX), 4mL DAB Plus Chromogen (TA-004-HCX)
Chromagen Diaminobenzidine (DAB)
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Storage Requirements Store at 2° to 8°C. Each component is stable for 18 months.
Enzyme Peroxidase
Product Line Lab Vision, UltraVision
Substrate Diaminobenzidine (DAB)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.