Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ UltraVision™ Quanto Detection System HRP

Catalog No. TL060QHL Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL060QHL 60 mL
TL125QHL 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. TL060QHL Supplier Epredia™ Supplier No. TL060QHL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Epredia™ UltraVision Quanto HRP DAB delivers higher lab profitability and better staining by supporting laboratories to select the best antibody for each test.

UltraVision Quanto is the technology of this decade in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This innovative micropolymer technology has major advantages over conventional IHC systems as it allows the “detector” to readily penetrate to the nuclear membrane enabling the user to detect nuclear antigens without sacrificing the ability to stain cytoplasmic and membrane antigens. UltraVision Quanto amplifies the signal with both mouse and rabbit primary antibodies making it versatile and easy to use. Background noise due to nonspecific binding to endogenous biotin molecules is eliminated because UltraVision Quanto is not a biotin/avidin based system. The signal to noise ratio is further enhanced to detect low antigen expression or compensate for low primary antibody affinity by utilizing highly purified secondary antibodies and enzymes. The UltraVision Quanto ultimately provides the user with a rapid, easy to use, and versatile IHC detection system that offers superior staining of tissues traditionally thought difficult to visualize.
  • Enhanced sensitivity while minimizing background — Can be used with low expression and routine markers alike
  • Increased sensitivity for rabbit antibodies — more economical use of expensive rabbit monoclonal antibodies
  • Reduced cost per test — Quanto remains one of the most affordable systems available
  • Shorter incubation times — Faster turn around times
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes: TL-060-HL: 60mL Primary Antibody Amplifier Quanto (TL-060-QPB), 60mL HRP Polymer Quanto (TL-060-QPH), 60mL Ultra V Block (TA-060-UB)

TL-125-HL: 125mL Primary Antibody Amplifier Quanto (TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB),

Specifications

Quantity 60 mL
Includes 60mL Primary Antibody Amplifier Quanto (TL-060-QPB), 60mL HRP Polymer Quanto (TL-060-QPH), 60mL Ultra V Block (TA-060-UB)
Chromagen None provided
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Storage Requirements Store at 2° to 8°C. Each component is stable for up to 18 months.
Enzyme Peroxidase
Product Line UltraVision
Substrate None provided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.