Learn More
Description
- Enhanced sensitivity while minimizing background — Can be used with low expression and routine markers alike
- Increased sensitivity for rabbit antibodies — more economical use of expensive rabbit monoclonal antibodies
- Reduced cost per test — Quanto remains one of the most affordable systems available
- Shorter incubation times — Faster turn around times
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: None provided
Includes: TL-060-HL: 60mL Primary Antibody Amplifier Quanto (TL-060-QPB), 60mL HRP Polymer Quanto (TL-060-QPH), 60mL Ultra V Block (TA-060-UB)
TL-125-HL: 125mL Primary Antibody Amplifier Quanto (TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB),
Specifications
Specifications
| Includes | 125mL Primary Antibody Amplifier Quanto (TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB) |
| Chromagen | None provided |
| Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
| Storage Requirements | Store at 2° to 8°C. Each component is stable for up to 18 months. |
| Enzyme | Peroxidase |
| Substrate | None provided |
| Quantity | 125 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.