Learn More
Electron Microscopy Sciences EMS Glass Knife Strip Ultramicrotomy Grade, 12 mm thick, 50 x 100 mm
SDP
Catalog No. 50930673
Encompass_Preferred
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 12 mm
Size: 50 x 100 mm
Catalog No. 50-930-673 Supplier Electron Microscopy Sciences Supplier No. 71015
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 12 mm
Size: 50 x 100 mm