Learn More
Crescent Chemical Co Inc Ultroser G Serum 20mL
SDP
Catalog No. NC1700979
Encompass_Preferred
Ultroser™ G serum substitute can successfully replace FCS (fetal calf serum) in cell culture medium. It is designed for the in vitro culture of anchorage-dependent cells. It is specially tested as the primary culture from amniotic liquid cells and is designed to confirm their ability for karyotyping.
Catalog No. NC1700979 Supplier Crescent Chemical Co Inc Supplier No. 67042.1EA
Ultroser™ G serum substitute can successfully replace FCS (fetal calf serum) in cell culture medium. It is designed for the in vitro culture of anchorage-dependent cells. It is specially tested as the primary culture from amniotic liquid cells and is designed to confirm their ability for karyotyping.