Learn More
Hunter Lab UltraScan VIS Spectrophotometer
SDP
Catalog No. NC2005792
Encompass
The UltraScan VIS color measurement spectrophotometer measures opaque, transparent, and translucent solids and liquids in the complete CIE recommended spectral range of 360-780 nm. It can be used in production and in the laboratory for inspecting raw materials, evaluating finished product or developing methods for color analysis.
Catalog No. NC2005792 Supplier Hunter Lab Supplier No. USVIS
Add to Cart
Add to Cart
Request a Quote
The UltraScan VIS color measurement spectrophotometer measures opaque, transparent, and translucent solids and liquids in the complete CIE recommended spectral range of 360-780 nm. It can be used in production and in the laboratory for inspecting raw materials, evaluating finished product or developing methods for color analysis.