Learn More
Electron Microscopy Sciences DiATOME Ultra-Semi Diamond Knife 3.0 mm 35° Knife angle
SDP
Catalog No. 501902719
Encompass
Similar to our ultra 35° 3.0 mm with a greater thickness range (50-500nm)
Size: 3.0 mm
Knife angle: 35°
Catalog No. 50-190-2719 Supplier Electron Microscopy Sciences Supplier No. 30SL
Similar to our ultra 35° 3.0 mm with a greater thickness range (50-500nm)
Size: 3.0 mm
Knife angle: 35°