Learn More
ORS Nasco ULTRSPEC 2001DUO TEM CL UD1PR
Catalog No. 502551592
Encompass_Preferred
Honeywell part# S0112. Honeywell Uvex Ultraspec 2001 OTG Safety Glasses Lens Type: Scratch-resistant Lens Tint: Clear Lens Coating: Ultra-dura Hardcoat UV Protection: 99.90% Certifications/Compliance: ANSI Z87.1-2015 CSA Z94.3-2010 Eyewear Recommended Applications: Safety Temple Style: Duoflex Weight: 51g
Catalog No. 50-255-1592 Supplier ORS Nasco Supplier No. 763S0112
Honeywell part# S0112. Honeywell Uvex Ultraspec 2001 OTG Safety Glasses Lens Type: Scratch-resistant Lens Tint: Clear Lens Coating: Ultra-dura Hardcoat UV Protection: 99.90% Certifications/Compliance: ANSI Z87.1-2015 CSA Z94.3-2010 Eyewear Recommended Applications: Safety Temple Style: Duoflex Weight: 51g