Learn More
Azzota Corp 8.5mm Window Height, Ultra-Micro Uv-Transparent Disposable Cuvette, Dimensions 12.5mm Width X 12.5mm Depth X 45.0mm Height, 10mm Light Path, Filling Volume 70L To 550L, Window D 2X3.5mm, 100/Pk
Catalog No. 502060338
Encompass_Preferred
8.5mm window height, Ultra-micro UV-Transparent Disposable Cuvettes, Cuvette Dimensions 12.5mm width x 12.5mm Depth x 45.0mm height, 10mm Light Path, Filling Volume 70L to 550L, Window Dimensions 2x3.5mm, 100/pk
Catalog No. 50-206-0338 Supplier Azzota Corp Supplier No. A906
8.5mm window height, Ultra-micro UV-Transparent Disposable Cuvettes, Cuvette Dimensions 12.5mm width x 12.5mm Depth x 45.0mm height, 10mm Light Path, Filling Volume 70L to 550L, Window Dimensions 2x3.5mm, 100/pk