Learn More
LifeSensors Tandem Ubiquitin Binding Entities (TUBE2) HRP 50µg
SDP
Catalog No. NC2383157
Encompass_Preferred
Based on protein domains known to possess an affinity for ubiquitin, Tandem Ubiquitin Binding Entities (TUBEs) have been developed for the isolation and identification of ubiquitylated proteins. TUBEs display up to a 1000-fold increase in affinity for polyubiquitin moieties over the single ubiquitin binding associated domain (UBA). In addition, TUBEs display a protective effect on polyubiquitinated proteins, allowing for detection at relatively low abundance. These properties effectively “capture” proteins in their polyubiquitylated state. We have developed the HRP-TUBE 2 for quick and easy detection of polyubiquitin and polyubiquitinated substrates. This product is protected by one or more US or Foreign patents.
Catalog No. NC2383157 Supplier LifeSensors Supplier No. UM03120050
Based on protein domains known to possess an affinity for ubiquitin, Tandem Ubiquitin Binding Entities (TUBEs) have been developed for the isolation and identification of ubiquitylated proteins. TUBEs display up to a 1000-fold increase in affinity for polyubiquitin moieties over the single ubiquitin binding associated domain (UBA). In addition, TUBEs display a protective effect on polyubiquitinated proteins, allowing for detection at relatively low abundance. These properties effectively “capture” proteins in their polyubiquitylated state. We have developed the HRP-TUBE 2 for quick and easy detection of polyubiquitin and polyubiquitinated substrates. This product is protected by one or more US or Foreign patents.