Learn More
LifeSensors Tandem Ubiquitin Binding Entities (TUBE2) TAMRA 50µg
SDP
Catalog No. NC2060180
Encompass_Preferred
Based on protein domains known to possess an affinity for ubiquitin, Tandem Ubiquitin Binding Entities (TUBEs) have been developed for the isolation and identification of ubiquitylated proteins. TUBEs display up to a 1000-fold increase in affinity for poly-ubiquitin moieties over the single ubiquitin binding associated domain (UBA). In addition, TUBEs display a protective effect on polyubiquitylated proteins, allowing for detection at relatively low abundance. These properties effectively “capture” protein in its polyubiquitin state. TAMRA-TUBE 2 has a single TAMRA fluorophore (Exc. = 540 nm, Emm. = 578 nm) attached to the fusion tag of TUBE 2 (Cat. No. UM202). Since the fluorophore is attached to the tag, it does not affect binding of the tandem ubiquitin binding domains to polyubiquitin chains. This product is protected by one or more US or Foreign patents.
Catalog No. NC2060180 Supplier LifeSensors Supplier No. UM0502T0050
Based on protein domains known to possess an affinity for ubiquitin, Tandem Ubiquitin Binding Entities (TUBEs) have been developed for the isolation and identification of ubiquitylated proteins. TUBEs display up to a 1000-fold increase in affinity for poly-ubiquitin moieties over the single ubiquitin binding associated domain (UBA). In addition, TUBEs display a protective effect on polyubiquitylated proteins, allowing for detection at relatively low abundance. These properties effectively “capture” protein in its polyubiquitin state. TAMRA-TUBE 2 has a single TAMRA fluorophore (Exc. = 540 nm, Emm. = 578 nm) attached to the fusion tag of TUBE 2 (Cat. No. UM202). Since the fluorophore is attached to the tag, it does not affect binding of the tandem ubiquitin binding domains to polyubiquitin chains. This product is protected by one or more US or Foreign patents.